Learn More
Cayman Chemical ProcarbazInhydrochlorIde 1g
SDP
Supplier:  Cayman Chemical 161331 G
Encompass_Preferred
A methyl hydrazine derivative that is converted by metabolism into a methylating species which converts DNA to various DNA adducts, including O6-methylguanine
Catalog No. 50-297-0351