Learn More
Medchemexpress LLC Prochloraz | 67747-09-5 | 99.85% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB08451G
Encompass_Preferred
Prochloraz is an imidazole antifungal that functions as an estrogen receptor (ER) and androgen receptor (AR) antagonist, and an aromatase inhibitor. It strongly inhibits the microsomal cytochrome P-450 dependent 14α-demethylation of lanosterol during ergosterol biosynthesis, leading to membrane disruption and eventual cell death. In vivo, it acts as an antiandrogen by reducing reproductive organ weights, affecting androgen-regulated gene expression in the prostate, and increasing luteinizing hormone levels.
  • Acts as an estrogen receptor (ER) and androgen receptor (AR) antagonist
  • Functions as an aromatase inhibitor
  • Activates the aryl hydrocarbon receptor (AhR)
  • Inhibits ergosterol biosynthesis
  • Causes membrane disruption and cell death
  • Reduces reproductive organ weights in vivo
  • Affects androgen-regulated gene expressions in the prostate
  • Increases luteinizing hormone levels
Catalog No. 50-004-07948