Learn More
Medchemexpress LLC Prochloraz manganese | 75747-77-2 | 1632.51 | C60H64Cl14MnN12O8 | 5 MG

Supplier: Medchemexpress LLC HY1373565MG
Prochloraz manganese is the manganese salt form of the fungicide prochloraz provided as an analytical standard for research and environmental/agricultural analysis. It is intended for laboratory use as a reference material in method development, residue analysis, and quality control.
- Antifungal agent used in agricultural and environmental analysis.
- Provided as an analytical/research standard for laboratory applications.
- Available in small vial pack sizes suitable for trace-level analyses.
- Chemical formula C60H64Cl14MnN12O8.
- Molecular weight 1632.51.
- Intended for method development, residue testing, and quality control workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.