Learn More
Medchemexpress LLC Prochloraz manganese | 75747-77-2 | C60H64Cl14MnN12O8 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13735610MG
Encompass_Preferred
Prochloraz manganese is the manganese complex of the fungicide prochloraz supplied as a research/analytical standard. It is used in agricultural and laboratory studies as an antifungal reference material and is intended for research use only.
  • Antifungal agent that inhibits sterol C-14 demethylation.
  • Provided as an analytical reference standard for laboratory and agricultural research.
  • Characterized by molecular formula C60H64Cl14MnN12O8 and molecular weight 1,632.51 g/mol.
  • Available in small, pre-weighed packages suitable for analytical workflows.
  • Intended for research use only; not for human or veterinary applications.
Catalog No. 50-002-59629