Learn More
Lonza Walkersville ProCHO 5 CHO Med Powder 500L
Supplier:  Lonza Walkersville WPW045L
Encompass
ProCHO 5 Powder Kits (500L) protein-free CHO Media were developed specifically to facilitate the production and downstream processing of recombinant proteins expressed in CHO cells These protein-free formulations support high-density cultures without the need for animal derived components Very low levels of human recombinant insulin facilitate both downstream purification and regulatory compliance ProCHO 5 - For CHO cells already growing in suspension Supports increased protein production Contains 0 1% Pluronic F-68 Does not contain L-glutamine phenol red hypoxanthine or thymidine ProCHO 5 basal powder allow to produce liquid medium in any required volume make sure to include the required Insulin and lipids This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only
Catalog No. 50-004-96214