Learn More
Lonza Walkersville ProCHO 4 Protein-free CHO Medium, 1L
Supplier:  Lonza Walkersville BEBP12029Q
Encompass
ProCHO 4 Protein-free CHO Medium is developed specifically to facilitate the production and downstream processing of recombinant proteins expressed in CHO cells. These protein-free formulations support high-density cultures without the need for animal derived components. Very low levels of recombinant insulin facilitate both downstream purification and regulatory compliance.ProCHO 4 Protein-free CHO Medium for concurrent transition of adherent CHO cells to serum-free and suspension culture. Supports faster doubling times.Benefits:ProCHO 4 Protein-free CHO MediumSerum-free cell culturingProtein Free MediumNon-animal originApplications:Protein production in CHO cells
Catalog No. 50-212-0213