Learn More
Enterprise Technology Solutions Procter & Gamble Commercial Air Refresher, Linen and Sky Scent, 8.8 oz., Multi
SDP
Supplier:  Enterprise Technology Solutions PGC96256
Encompass_Preferred
Air refresher features OdorClear technology that eliminates lingering odors on the spot to leave rooms with nothing but a light, fresh scent. Its special odor neutralizers bind to airborne malodors to reduce and neutralize their pH value for complete freshness. Linen and Sky scent lets you bring the outdoors in with a fresh off the clothesline scent. Natural propellant delivers a continuous flow of freshness without VOCs for safe use. Air refresher is perfect for clearing away odors from everyday pet and bathroom smells to stubborn smoke and leftover cooking odors.
Catalog No. 50-007-90452