Learn More
Enterprise Technology Solutions Procter & Gamble Commercial FabricSpray,Antibacterial,Deodorizer,LightScent,22oz,OE
SDP
Supplier:  Enterprise Technology Solutions PGC76533
Encompass_Preferred
Tide Antibacterial Fabric Spray is specially designed to kill 99.9% of bacteria left behind on fabrics and fight germs on hard surfaces (when used as directed). Every day, we come in contact with a variety of unclean surfaces, and millions of bacteria from these surfaces can linger on our clothes, linens, and upholstery. Lucky for you, Tide's sanitizing spray keeps your home germ-free and fresh by eliminating those pesky bacteria from hard-to-wash-items. Tide Antibacterial Fabric Spray is also a worthy solution for anyone looking to eliminate odors and refresh clothes in between washes. So, when the thought of bacteria crawling on your backpack makes you shudder, just spray it, don't say it. Antibacterial spray is effective against Staphylococcus aureus, Salmonella enterica, Escherichia coli and Proteus mirabilis on hard, non-porous surfaces, and it's effective against Enterobacter aerogenes, Staphylococcus aureus on soft surfaces/fabrics (when used as directed).
Catalog No. 50-007-90704