Learn More
TARGETMOL CHEMICALS INC Procyanidin A1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN11365MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes.Procyanidin A1 (Proanthocyanidin A1) has antiallergic effects, it inhibits degranulation downstream of protein kinase C activation or Ca2+; influx from an internal store in RBL-2H3 cells. Purity 99.23%
Catalog No. 50-287-7227