Learn More
AdipoGen Procyanidin A2,Chemical. CAS: 41743-41-3
SDP
Supplier:  AdipoGen AGCN20486M001
Encompass_Preferred
Procyanidin A2, AdipoGen Life Sciences. Chemical. CAS: 41743-41-3. Formula: C30H24O12. MW: 576.5. Isolated from grape seeds (Vitis vinifera L). A-Type proanthocyanidine. Potent antioxidant. Free radical scavenger and lipid peroxidation inhibitor. Potent low-density lipoprotein (LDL) oxidation inhibitor. Antidiabetic. Prevents islet cell apoptosis, hyperglycemia and type 2 diabetes through it's antioxidant properties. Antibacterial agent. Antiviral. Shown to inhibit viral replication of canine distemper virus (CDV). Glycosyltransferase B (GTFB) inhibitor. 41743-41-3
Catalog No. 50-117-6035