Learn More
TARGETMOL CHEMICALS INC Procyanidin A2 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN11325MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Procyanidin A2 is a potential precursor of 5-(3',4'-dihydroxyphenyl)-gamma-valerolactone, exhibits antioxidant, anti-inflammary, anti-hyperglycemia, and anti-type 2 diabetes activities. Procyanidin A2 demonstrates liver cell regenerative activity in scratch wound healing assays and alcohol-induced liver cell injury in vitro. Purity 98.78%
Catalog No. 50-288-0175