Learn More
Medchemexpress LLC HY-N2342 1mg Medchemexpress, Procyanidin C1 CAS:37064-30-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN23421MG
Encompass_Preferred
Medchemexpress, HY-N2342 1mg Procyanidin C1 CAS:37064-30-5 Procyanidin C1 is a natural polyphenol, causes DNA damage, cell cycle arrest, and induces apoptosis. Procyanidin C1 decreases the level of Bcl-2, but enhances BAX, caspase 3 and 9 expression in cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0384