Learn More
Alkali Scientific ProCycle™ 12-Strip Caps for 1.2 mL Cluster (Dilution) Tubes, 80 Strips/Pk, Non-Sterile
SDP
Supplier:  Alkali Scientific CTC1212
Encompass_Preferred
Strip caps are compatible with CT1212 12-strip tubes. Ideal for automation / HTS. Strip caps are polyethylene. The tube-and-rack systems are suitable as a reaction vessel in your robotic workstation or as a long-term storage vessel in your sample library. Certified free of detectable RNase DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. 50-302-8597