Learn More
Alkali Scientific ProCycle™ Pre-cut Sealing Membranes, Non-Sterile, qPCR Optical (pressure-sensitive adhesive) ABI™ Type, 10 bags of 10 membranes
SDP
Supplier:  Alkali Scientific SF1001QPS
Encompass
RNase, DNase, DNA, and PCR inhibitor free Evaporation rates ≤ 5% Inner packs of 10 membranes PureAmp™ Sealing Membranes are compatible with all standard PCR plates. Membrane options include pierceable aluminum, standard transparent, optically clear sticky adhesive, optically clear pressure-sensitive adhesive, and low-adhesive EZ-peel, all of which are guaranteed to keep evaporation rates below 5%.
Catalog No. 50-302-6913