Learn More
Cayman Chemical ProcynIdIn A2 10mg
SDP
Supplier:  Cayman Chemical 2520010 MG
Encompass_Preferred
A flavanol dimer with diverse biological properties; scavenges DPPH radicals (IC50 = 5.08 μM); inhibits PDGF-induced STAT3 activation in primary rat VSMCs at 30 μg/ml; inhibits growth of S. aureus and E. coli (MICs = 62.5 and 62.5 μg/ml, respectively) and proliferation of human HepG2 liver hepatocellular carcinoma and HeLa cervical cancer cells (EC50s = 62.19 and 66.07 μg/ml, respectively); increases insulin secretion from primary mouse pancreatic islets at 10 μM; inhibits bisphenol A-induced glucose increases in fasted mice at 10 μmol/kg per day
Catalog No. 50-293-2621