Learn More
Cayman Chemical ProcynIdIn B4 10mg
SDP
Supplier:  Cayman Chemical 3639810 MG
Encompass_Preferred
An ACSL4 inhibitor selective for ACSL4 over ACSL1 and ACSL3 inhibits formation of AA-CoA from arachidonic acid in as well as the proliferation and migration of PC3 and MDA-MB-231 cancer cells inhibits tumor growth in a PC3 mouse xenograft model at 0 25 mg/kg
Catalog No. 50-291-2142