Learn More
Apexbio Technology LLC Prodan 70504-01-7 25mg
SDP
Supplier:  Apexbio Technology LLC A471825
Encompass_Preferred
Prodan (CAS 70504-01-7) is a fluorescent dye utilized as an environment-sensitive probe in bio-membrane studies It enables analysis of membrane dynamics by exhibiting emission spectrum shifts in response to changes in local solvent polarity and membrane micro-environments Prodan exerts its biological activity primarily by functioning as a fluorophore sensitive to solvent orientation polarizability facilitating investigation of lipid organization membrane fluidity and protein interactions Modifications of Prodan have also enabled the synthesis of fluorescent nucleoside analogs for the study of cellular signaling and nucleic acid interactions Based on these properties Prodan holds research potential in cellular biology for elucidating membrane organization and signaling mechanisms
Catalog No. 50-000-95097