Learn More
Medchemexpress LLC Prodan | 70504-01-7 | 99.7% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYD007450MG
Encompass_Preferred
Prodan is a solvatochromic fluorophore used as a microenvironment-sensitive membrane reporter. Fluorescent nucleosides designed and synthesized based on Prodan's chemistry have shown sensitive variations in Stokes shift values depending on the solvent's orientational polarizability.
  • Solvatochromic fluorophore.
  • Microenvironment-sensitive membrane reporter.
  • Used in the design and synthesis of fluorescent nucleosides that sensitively vary Stokes shift values based on solvent orientational polarizability.
Catalog No. 50-004-10145