Learn More
Selleck Chemical LLC Proflavine Hemisulfate S4236-1g
SDP
Supplier:  Selleck Chemical LLC S42361G
Encompass_Preferred
Proflavine Hemisulfate is a topical antiseptic by interchelating DNA thereby disrupting DNA synthesis and leading to high levels of mutation in the copied DNA strands
Catalog No. 50-311-1198