Learn More
Medchemexpress LLC Progabide | 62666-20-0 | 98.5% | C17H16ClFN2O2 | 100 MG

Supplier: Medchemexpress LLC HYA0173100MG
Progabide is a gamma-aminobutyric acid receptor (GABA) agonist, available for research use. It is a well-characterized compound with a purity of 98.5%.
- GABA receptor agonist
- For research use only
- High purity of 98.5%
- Stable as a powder for up to 3 years at -20°C, or 2 years at 4°C
- Stable in solvent for up to 6 months at -80°C, or 1 month at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.