Learn More
Medchemexpress LLC Progabide | 62666-20-0 | 98.5% | C17H16ClFN2O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYA0173
Encompass_Preferred
Progabide is a gamma-aminobutyric acid receptor (GABA) agonist. This compound is intended for research use only.
  • Acts as a GABA receptor agonist.
  • Can increase plasma corticosterone levels in rats, suitable for in vivo studies.
  • High purity of 98.5%.
  • Available in powder form for preparing solutions.
  • Offers long-term stability with proper storage.
Catalog No. 50-003-57676