Learn More
Medchemexpress LLC Progabide | 62666-20-0 | 98.5% | C17H16ClFN2O2 | 5 MG

Supplier: Medchemexpress LLC HYA01735MG
Progabide is a gamma-aminobutyric acid receptor (GABA) agonist. It has been shown to increase plasma corticosterone levels in male Wistar rats in a dose-dependent manner.
- Acts as a GABA receptor agonist.
- Increases plasma corticosterone levels in male Wistar rats.
- Molecular weight: 334.77.
- Appearance: Solid.
- Color: Light yellow to yellow.
- Soluble in DMSO (250 mg/mL).
- Store powder at -20°C for 3 years, or 4°C for 2 years.
- Store in solvent at -80°C for 6 months, or -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.