Learn More
Medchemexpress LLC Progabide | 62666-20-0 | 98.5% | C17H16ClFN2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA017350MG
Encompass_Preferred
Progabide is a gamma-aminobutyric acid receptor (GABA) agonist. It is intended for research use only and is not sold to patients. It is a solid with a light yellow to yellow color and a molecular weight of 334.77.
  • Functions as a GABA receptor agonist.
  • Soluble in DMSO at 250 mg/mL (746.78 mM) with sonication.
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month.
  • Increases plasma corticosterone levels in male Wistar rats in vivo.
  • Associated with research in neurological, eye or ear diseases, and neurodegenerative diseases.
  • Relevant to various compound screening libraries for neuronal signaling and GABA receptors.
Catalog No. 50-004-32504