Learn More
American Research Products Inc AAV2 Xpress ELISA | 96 tests
Supplier:  American Research Products Inc PRAAV2XP
Encompass
Fast, easy and reliable AAV2 titer determination with PROGEN's AAV2 Xpress ELISA: assay time was reduced to less than 2 hours by adjustment of the kit components and the protocol of our standard AAV2 Titration ELISA 2.0R, seamless and risk-free transition from our standard AAV2 Titration ELISAs (PRAAV2R and PRATV) to the Xpress version since the composition of the kit’s core characteristics are not changed, same accuracy and reproducibility as the standard AAV2 Titration ELISAs.
Catalog No. NC1858541