Learn More
Cayman Chemical Progesteron ELISA Stndard 1ea
SDP
Supplier:  Cayman Chemical 4826041 EA
Encompass_Preferred
A phospholipid has been used in the generation of LNPs for the delivery of mRNA in vivo has been used in the synthesis of various PEG-conjUGates for incorporation into liposomes LNPs containing 12-dierucoyl-sn-glycero-3-PE and encapsulating mRNA encoding hEPO increase serum hEPO levels in mice
Catalog No. 50-289-6836