Learn More
Medchemexpress LLC Progesterone-d9 (Pregn-4-ene-3,20-dione-d9) | 15775-74-3 | 323.52 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0437S5MG
Encompass_Preferred
Progesterone-d9 is a deuterium-labeled version of Progesterone, a steroid hormone essential for regulating the menstrual cycle and crucial for pregnancy.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89800