Learn More
Arbor Assays Progesterone ELISA Kit (1 Strip Plate)
SDP
Supplier:  Arbor Assays K025H1
Encompass_Preferred
The DetectX Progesterone ELISA Kits quantitatively measure Progesterone present in extracted dried fecal samples, urine and tissue culture media samples. Used in determination of Reproduction and Sex Steroid Metabolism
Catalog No. 50-227-8424