Learn More
IBL America Progesterone (Mouse/Rat) ELISA
Supplier:  IBL America IB79183
Encompass_Preferred
Competitive enzyme immunoassay (ELISA) for the determination of progesterone in rat and mouse serum or plasma. For research use only, not for use in diagnostic procedures.An unknown amount ofprogesterone present in the sample and a defined amount of progesterone conjugated to horseradishperoxidase compete for the binding sites of progesterone antiserum coated to the wells of a microplate.After incubation on a shaker the microplate is washed four times. After addition of the substrate solutionthe concentration of progesterone is inversely proportional to the optical density measured.
Catalog No. 50-153-8196