Learn More
Medchemexpress LLC Progonadoliberin-2 (human recombinant, His-tag) | 50UG
SDP
Supplier:  Medchemexpress LLC HYP7020550UG
Encompass_Preferred
Recombinant human progonadoliberin-2 (GnRH II) produced in HEK293 cells with a His-tag. Supplied as a lyophilized powder (50 μg), this protein includes the full amino acid sequence and is provided with a low endotoxin specification for research applications involving endocrine signaling and gonadotropin secretion. Follow storage and reconstitution recommendations for best performance.
  • Lyophilized powder for long-term storage.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • His-tagged to facilitate purification and detection.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Contains full amino acid sequence for complete structural studies.
  • Stable when aliquoted and frozen according to supplier instructions.
Catalog No. 50-001-76276