Learn More
Medchemexpress LLC Proguanil | 500-92-5 | 99.84% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB080610MM/1ML
Encompass_Preferred
Proguanil is an antimalarial proagent that is metabolized into its active form, Cycloguanil. It functions as a potent inhibitor of dihydrofolate reductase (DHFR). This compound is intended for research purposes only and is not suitable for patient use.
  • Antimalarial proagent
  • Metabolized to active form, Cycloguanil
  • Dihydrofolate reductase (DHFR) inhibitor
  • For research use only
Catalog No. 50-004-09468