Learn More
Selleck Chemical LLC Proguanil HCl(Atovaquone) S3082-10mg
SDP
Supplier:  Selleck Chemical LLC S308210MG
Encompass_Preferred
Proguanil hydrochloride(Chloroguanil Chlorguanide hydrochloride) an antimalarial proagent acts as an inhibitor of dihydrofolate reductase (DHFR) Proguanil acts as a biguanide to enhance the Atovaquone effect
Catalog No. 50-311-1584