Learn More
TARGETMOL CHEMICALS INC Proguanil hydrochloride 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T438225MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Proguanil hydrochloride (Chloroquanil) is a biguanide compound which metabolizes in the body to form cycloguanil, an anti-malaria agent.Upon hydrolysis, Proguanil hydrochloride is converted to its active cyclic triazine metabolite, cycloguanil, by a cytochrome P450 dependent reaction. Cycloguanil selectively inhibits the bifunctional dihydrofolate reductase-thymidylate synthase of plasmodium parasite, thereby disrupting deoxythymidylate synthesis and ultimately blocking DNA and protein synthesis in the parasite. Purity 98.13%
Catalog No. 50-287-8894