Learn More
Medchemexpress LLC Proguanil (hydrochloride) | 637-32-1 | 99.2% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0806A100MG
Encompass_Preferred
Proguanil hydrochloride is an antimalarial proagent that is metabolized into its active metabolite, Cycloguanil. It acts as a dihydrofolate reductase (DHFR) inhibitor, primarily used in research to study malaria and its related pathways.
  • Functions as a dihydrofolate reductase (DHFR) inhibitor
  • Metabolized into active form Cycloguanil for antimalarial activity
  • Synergistic effects when combined with Atovaquone
  • Active against gametocytes and pre-erythrocytic stages of malaria parasites
  • Reversibly inhibits 5-HT3 receptor responses
  • Intended for research use only
Catalog No. 50-004-09469