Learn More
Medchemexpress LLC Proguanil hydrochloride | 637-32-1 | MFCD01732193 | 99.2% | 290.19 g·mol⁻¹ | C11H17Cl2N5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0806A10MG
Encompass_Preferred
Proguanil hydrochloride is an antimalarial prodrug that is metabolized to cycloguanil and acts as a dihydrofolate reductase (DHFR) inhibitor. It is supplied as a white to off-white solid for research use, with high purity and solvent solubility suitable for in vitro testing.
  • Antimalarial prodrug converted to cycloguanil.
  • Dihydrofolate reductase (DHFR) inhibitor used in antiparasitic research.
  • High purity suitable for analytical and biological assays.
  • Soluble in DMSO at high concentration; sonication may be required.
  • Available in small research pack sizes for convenience.
Catalog No. 50-002-97428