Learn More
Medchemexpress LLC Prohexadione calcium | 127277-53-6 | 98.1% | 251.27 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY112105
Encompass_Preferred
Prohexadione calcium (BX-112) is a plant growth regulator that inhibits gibberellin biosynthesis in plants. This product is for research use only and not sold to patients.
  • Inhibits gibberellin biosynthesis in plants
  • Inhibits JMJD2A demethylase activity at 1 mM
  • Induces the differentiation of mouse neurospheres into neurons (at 1-2 mM concentration)
  • Alleviates saline-alkali stress in soybean seedlings when sprayed at 100 mg/L
  • Induces structural resistance to fire blight infection in potted apple trees
Catalog No. 50-004-39517