Learn More
Essendant Epson PowerLite 1780W Wireless WXGA 3LCD Projector, 3,000 lm, 1280 x 800 Pixels, 1.2x Zoom
Supplier:  Essendant EPSV11H795020
Encompass_Preferred
Achieve brilliant presentations wherever business takes you with the ultra slim PowerLite 1780W wireless widescreen projector This thin 4 lb model delivers 3 000 lumens of color brightness and 3 000 lumens of white brightness1 plus WXGA resolution ideal for HD-quality content Offering 3x Higher Color Brightness2 than competitive models Epson 3LCD projectors ensure vivid presentations Featuring the latest in connectivity this projector makes it easy to wirelessly share content from your tablet or smartphone3 Or share HD videos using your streaming device The 1780W supports Chromecast Roku and MHL-enabled devices And it offers fast wireless setup with its on-screen QR code Brightness- 3 000 lm Technology- 3LCD Zoom- 1 2x Weight- 4 lbs
Catalog No. 50-000-35602