Learn More
Eurofins Discoverx PathHunter PROKR1 Total GPCR Internalization U2OS Cell Line, 2 vials. (93-0764C3)
Supplier:  Eurofins Discoverx 930764C3
Encompass
The PathHunter PROKR1 Total GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of PROKR1 via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged PROKR1 and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the PROKR1 receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1272
Add to Cart
Request a Quote