Learn More
Medchemexpress LLC Prolactin Human HE 100ug

Supplier: Medchemexpress LLC HYP71059100UG
Recombinant human prolactin expressed in HEK293 cells and supplied as a lyophilized, C-terminal His-tagged protein for research applications. The product is glycosylated and highly pure, with low endotoxin and recommended storage and formulation guidelines to preserve activity.
- Mammalian-expressed glycosylated protein suitable for functional studies.
- High purity (>95%) by reducing SDS-PAGE.
- Low endotoxin level (<1 EU/μg) for sensitive assays.
- Lyophilized powder formulation for long-term storage.
- Recommended storage at -20°C or -80°C with carrier protein for extended stability.
- Includes amino acid sequence and defined residue range for reproducibility.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.