Learn More
Medchemexpress LLC Prolactin Releasing Peptide (1-31), human acetate | 98.3% | 3724.17 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP1520A5MG
Encompass_Preferred
Prolactin Releasing Peptide (1-31), human (acetate) is a high affinity GPR10 ligand that causes the release of prolactin. It binds to GPR10 in humans and rats with Ki values of 1.03 nM and 0.33 nM, respectively. This peptide can be used for research related to the hypothalamo-pituitary axis.
  • High affinity GPR10 ligand
  • Causes the release of prolactin
  • Binds to human GPR10 with a Ki of 1.03 nM
  • Binds to rat GPR10 with a Ki of 0.33 nM
  • Used for research of the hypothalamo-pituitary axis
  • Increases plasma FSH and total plasma testosterone
  • Increases LHRH release from hypothalamic explants
  • Increases hypothalamic peptides (VIP and galanin) involved in pituitary hormone release
Catalog No. 50-004-24312