Learn More
CPC Scientific H-Ser-Arg-Thr-His-Arg-His-Ser-Met-Glu-Ile-Arg-Thr-Pro-Asp-Ile-Asn-Pro-Ala-Trp-Tyr-Ala-Ser-Arg-Gly-Ile-Arg-Pro-Val-Gly-Arg-Phe-NH2 0.5MG
SDP
Supplier:  CPC Scientific PROL001A
Encompass
Multifunctional Peptide in NeuroendocrinologySEQUENCE: H-Ser-Arg-Thr-His-Arg-His-Ser-Met-Glu-Ile-Arg-Thr-Pro-Asp-Ile-Asn-Pro-Ala-Trp-Tyr-Ala-Ser-Arg-Gly-Ile-Arg-Pro-Val-Gly-Arg-Phe-NH2ONE-LETTER SEQUENCE: SRTHRHSMEIRTPDINPAWYASRGIRPVGRF-NH2MOLECULAR FORMULA: C160H252N56O42S1MOLECULAR WEIGHT: 3664.2STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [215510-22-8]SYNONYMS: PrRP31 (human), Preprolactin (23-53) (human), Prolactin-Releasing Peptide (1-31) (human)RESEARCH AREA: HormonalREFERENCES:Engstr?m, et al. J. Pharmacol. Exp. Ther. 305, 825 (2003) S. Hinuma, et al. Nature, 393, 272 (1998) F. Satoh, et. al. J. Pharmacol.,129, 1787 (2000)
SKU(s): PROL-001A, PROL-001B
Catalog No. 50-211-0327