Learn More
Biotium Proliferation Viability Assays Enzyme Substrates Luciferase Substrates CellTiter Glo 1/EA
Supplier:  Biotium 301383
Encompass_Preferred
A luciferase-based ATP cell viability assay for high-throughput luminescence detection in microplates The homogenous assay offers high sensitivity wide dynamic range of over 3-4 orders of magnitude and long-lasting signal with a half-life of over 5 hours
Catalog No. 50-005-01478