Learn More
American Radiolabeled Chemicals Inc PROLINE L-1-14C 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 357350UC
Encompass_Preferred
Proline, L-1-14C 50uCi in ethanol:water (2:98), 0.1 mCi/ml, 50-60 mCi/mmol, M.W. 115.1, Exclusive
Catalog No. 50-753-1981