Learn More
Medchemexpress LLC Promazine hydrochloride | 53-60-1 | 99.87% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1225500MG
Encompass_Preferred
Promazine (Romtiazin) hydrochloride is an antipsychotic and a dopamine receptor D2 antagonist. It inhibits dopaminergic neurotransmission.
  • Acts as a dopamine receptor D2 antagonist.
  • Inhibits dopaminergic neurotransmission.
  • Shows KDs of 25 nM at the human norepinephrine transporter.
  • Shows KDs of 190 nM at the serotonin transporter.
  • Shows KDs of 8400 nM at the dopamine transporter.
Catalog No. 50-004-12871