Learn More
Medchemexpress LLC Promazine hydrochloride | 53-60-1 | 99.9% | 100 MG

Supplier: Medchemexpress LLC HYB1225100MG
Promazine hydrochloride is an antipsychotic and a dopamine receptor D2 antagonist that inhibits dopaminergic neurotransmission.
- IC50 & Target: D2 Receptor
- Shows KDs of 25, 190, and 8400 nM at the human norepinephrine transporter, serotonin transporter, and dopamine transporter respectively in vitro
- Molecular weight: 320.88
- Formula: C17H21ClN2S
- CAS number: 53-60-1
- Appearance: Solid
- Color: White to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.