Learn More
Promega Luciferin-EF™ Endotoxin-Free Luciferin Na
Luciferin-EF is an endotoxin-free beetle luciferin that can be used for imaging applications where endotoxins may create problems in living systems.
Supplier:  Promega E6552
Encompass
Catalog No. PRE6552
Add to Cart
Description
Description
  • Produces no potential interference in assay due to the presence of endotoxins
  • Packaged in amber vials with septa to ensure product integrity and offer ease of dilution and use for imaging experiments
Product Suggestions
Product Suggestions