Learn More
Medchemexpress LLC Promegestone (R-5020) | 34184-77-5 | 99.1% | 5 MG

Supplier: Medchemexpress LLC HY1193845MG
Promegestone (R-5020) is a synthetic progestin and potent progesterone receptor agonist used as a research tool for studies of progesterone signaling, endocrine regulation, and cancer biology.
- Potent progesterone receptor (PR) agonist for receptor biology studies.
- Synthetic steroid with molecular formula C22H30O2 and molecular weight 326.47 g/mol.
- Supplied as a solid suitable for in vitro and in vivo research applications.
- Typical purity reported around 98.0% by supplier certificate of analysis.
- Useful as a pharmacological probe in endocrine and cancer research models.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.