Learn More
Medchemexpress LLC Promethazine-d4 hydrochloride | 1173018-74-0 | 95.4% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0781S5MG
Encompass_Preferred
Promethazine-d4 (hydrochloride) is the deuterium labeled Promethazine hydrochloride. It is a first-generation antihistamine, a strong antagonist of the H1 receptor, and a moderate mACh receptor antagonist, with moderate affinity for 5-HT2A, 5-HT2C, D2, and α1-adrenergic receptors.
  • Deuterium labeled compound
  • Strong H1 receptor antagonist
  • Moderate mACh receptor antagonist
  • Moderate affinity for 5-HT2A, 5-HT2C, D2, and α1-adrenergic receptors
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-09148