Learn More
Selleck Chemical LLC Pronase E E8154-100MG
SDP
Supplier:  Selleck Chemical LLC E8154100MG
Encompass_Preferred
Pronase E (Activity 7000 U/g) is a mixture of proteolytic enzymes that is obtained from Streptomyces griseus and could digest protein into individual amino acids
Catalog No. 50-000-43701