Learn More
Medchemexpress LLC Propacetamol hydroch 10mM 1mL | 66532-86-3 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY10829210MM 1ML
Encompass_Preferred
Propacetamol hydrochloride is an analgesic agent can be used in induction of acute liver injury Propacetamol hydrochloride acts function through CYP2E1 inactivation UGT1A1 activation and attenuation of oxidative stress Propacetamol hydrochloride also is a precursor form of paracetamol Propacetamol hydrochloride can be used in postoperative pain acute trauma and gastrointestinal disorders[1
Catalog No. 50-002-42351