Learn More
Selleck Chemical LLC Propacetamol hydrochloride S4867-25mg
SDP
Supplier:  Selleck Chemical LLC S486725MG
Encompass_Preferred
Propacetamol hydrochloride (PRO) is a water-soluble prodrug of paracetamol (PA) which can be parenterally administered as analgesic for the treatment of postoperative pain acute trauma and gastric and/or intestinal disorders
Catalog No. 50-311-2117